LIF069953127 Life Chemicals Inc.

N-[(thiophen-2-yl)methyl]pyridin-2-amine

  • MDL number MFCD03826740
  • Cas Number : 140-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart