LIF069953130 Life Chemicals Inc.

1-(benzenesulfonyl)piperazine

  • MDL number MFCD02182804
  • Cas Number : 14172-55-5
  • Molecular Weight : 226.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart