LIF069955528 Life Chemicals Inc.

2-[4-(2,2-Dimethylpropanoyl)piperazin-1-yl]ethanol hydrochloride

  • MDL number MFCD07362917
  • Cas Number : 1049700-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart