LIF069957973 Life Chemicals Inc.

6-Hydroxy-7-methyl-4-(chloromethyl)-2H-chromen-2-one

  • MDL number MFCD04167233
  • Cas Number : 845637-29-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart