LIF069958096 Life Chemicals Inc.

4-(Chloromethyl)-6-hydroxy-2H-chromen-2-one

  • MDL number MFCD04180739
  • Cas Number : 847367-12-8
  • Molecular Weight : 210.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(chloromethyl)-6-hydroxy-2H-chromen-2-one
Inchi Code 1S/C10H7ClO3/c11-5-6-3-10(13)14-9-2-1-7(12)4-8(6)9/h1-4,12H,5H2
Inchi Key WGJIFHRBMQMSMV-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart