LIF069958923 Life Chemicals Inc.

4-(Chloromethyl)-5-hydroxy-7-methyl-2H-chromen-2-one

  • MDL number MFCD06644488
  • Cas Number : 879927-52-3
  • Molecular Weight : 224.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart