LIF069958988 Life Chemicals Inc.

{[5-(2-Thienyl)-1,3,4-oxadiazol-2-yl]thio}acetic acid

  • MDL number MFCD02741525
  • Cas Number : 485334-94-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart