LIF069961075 Life Chemicals Inc.

(2-Oxo-1,2-dihydro-quinolin-4-yl)-acetic acid

  • MDL number MFCD06253872
  • Cas Number : 21298-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart