LIF069961147 Life Chemicals Inc.

6-Chloro-pyrimidine-4,5-diamine

  • MDL number MFCD00023270
  • Cas Number : 4316-98-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart