LIF069961231 Life Chemicals Inc.

3-Chloro-1H-pyrazolo[4,3-c]pyridine-7-carbaldehyde

  • MDL number MFCD05668045
  • Cas Number : 93546-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart