LIF087878911 Life Chemicals Inc.

N-(sec-Butyl)tetrahydrothiophen-3-amine 1,1-dioxide hydrochloride

  • MDL number MFCD07359201
  • Cas Number : 1052420-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart