LIF087878912 Life Chemicals Inc.

1-(2-Methoxy-phenyl)-5-methyl-1H-[1,2,3]triazole-4-carboxylic acid

  • MDL number MFCD07397771
  • Cas Number : 210159-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart