LIF087878913 Life Chemicals Inc.

[3-(1H-Tetrazol-1-yl)phenyl]amine hydrochloride

  • MDL number MFCD06687920
  • Cas Number : 436100-10-6
  • Molecular Weight : 197.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart