LIF087878924 Life Chemicals Inc.

1-(2-Ethyl-phenyl)-5-oxo-pyrrolidine-3-carboxylic acid

  • MDL number MFCD03834519
  • Cas Number : 63675-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart