LIF087878960 Life Chemicals Inc.

5-Methyl-1-phenyl-1H-[1,2,3]triazole-4-carboxylic acid

  • MDL number MFCD00100217
  • Cas Number : 20725-32-0
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart