LIF087880753 Life Chemicals Inc.

4-(2-Phenoxyethoxy)benzoic acid

  • MDL number MFCD00020364
  • Cas Number : 22219-63-2
  • Molecular Weight : 258.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(2-phenoxyethoxy)benzoic acid
Inchi Code 1S/C15H14O4/c16-15(17)12-6-8-14(9-7-12)19-11-10-18-13-4-2-1-3-5-13/h1-9H,10-11H2,(H,16,17)
Inchi Key MIFKBDZVXBJUFX-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart