LIF087882129 Life Chemicals Inc.

1-Benzyl-4-oxo-1,4-dihydro-quinoline-3-carboxylic acid ethyl ester

  • MDL number MFCD08141530
  • Cas Number : 53977-02-9
  • Molecular Weight : 307.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 1-benzyl-4-oxo-1,4-dihydro-3-quinolinecarboxylate
Inchi Code 1S/C19H17NO3/c1-2-23-19(22)16-13-20(12-14-8-4-3-5-9-14)17-11-7-6-10-15(17)18(16)21/h3-11,13H,2,12H2,1H3
Inchi Key RWZQZQCEKZIQHV-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart