LIF090216048 Life Chemicals Inc.

2-(3-methyl-5-oxo-4,5-dihydro-1H-pyrazol-4-yl)acetic acid

  • MDL number MFCD04170350
  • Cas Number : 845808-92-6
  • Molecular Weight : 156.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart