LIF090217089 Life Chemicals Inc.

4,5,6,7-tetrahydro-2,1-benzisoxazol-3-amine

  • MDL number MFCD06255135
  • Cas Number : 13054-47-2
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart