LIF090224810 Life Chemicals Inc.

3-(2-Methylphenyl)-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD05170020
  • Cas Number : 1140528-29-5
  • Molecular Weight : 202.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart