LIF090224874 Life Chemicals Inc.

3-(2-Methoxyphenyl)-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD03030186
  • Cas Number : 834868-54-1
  • Molecular Weight : 218.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart