LIF090225014 Life Chemicals Inc.

3-(4-Ethoxyphenyl)-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD03030170
  • Cas Number : 1197631-30-3
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart