LIF090232646 Life Chemicals Inc.

2-Methyl-1,3-benzothiazol-5-amine

  • MDL number MFCD00226291
  • Cas Number : 13382-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart