LIF090233289 Life Chemicals Inc.

5-(2-thienyl)isoxazole-3-carboxylic acid

  • MDL number MFCD04113955
  • Cas Number : 763109-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart