LIF090236728 Life Chemicals Inc.

(4,5-Dimethyl-6-oxo-1,6-dihydro-pyrimidin-2-ylsulfanyl)-acetic acid

  • MDL number MFCD03765769
  • Cas Number : 499132-65-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart