LIF100864889 Life Chemicals Inc.

6-Methoxy-2H-chromene-3-carbaldehyde

  • MDL number MFCD01662418
  • Cas Number : 57543-40-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart