LIF100864899 Life Chemicals Inc.

6-Methyl-1H-pyrazolo[3,4-b]pyridin-3-amine

  • MDL number MFCD00456552
  • Cas Number : 79173-38-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart