LIF100864901 Life Chemicals Inc.

5-Hydroxy-1,2-dimethyl-1H-indole-3-carboxylic acid

  • MDL number MFCD00458839
  • Cas Number : 25888-01-1
  • Molecular Weight : 205.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart