LIF100864907 Life Chemicals Inc.

3,5-Dimethyl-1-(2-methylphenyl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD07186415
  • Cas Number : 400757-04-2
  • Molecular Weight : 214.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart