LIF100864914 Life Chemicals Inc.

1-(2-chlorobenzyl)piperazine hydrochloride

  • MDL number MFCD05656274
  • Cas Number : 856840-37-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart