LIF100864915 Life Chemicals Inc.

4,5,6,7,8,9-Hexahydro-1H-cycloocta[c]pyrazole-3-carboxylic acid

  • MDL number MFCD03452613
  • Cas Number : 650603-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart