LIF100868639 Life Chemicals Inc.

1-Benzoxepine-4-carboxylic acid

  • MDL number MFCD08541457
  • Cas Number : 923169-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart