LIF100870411 Life Chemicals Inc.

3-tert-butyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD00462163
  • Cas Number : 83405-71-4
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart