LIF100870431 Life Chemicals Inc.

3-(4,6-Dimethyl-2-oxopyrimidin-1(2H)-yl)propanoic acid hydrochloride

  • MDL number MFCD00449304
  • Cas Number : 463929-54-6
  • Molecular Weight : 232.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart