LIF100873139 Life Chemicals Inc.

1,2,5,6,7,8-hexahydroquinoline-2,5-dione

  • MDL number MFCD00766821
  • Cas Number : 15450-69-8
  • Molecular Weight : 163.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart