LIF100873165 Life Chemicals Inc.

2-Chloro-7,8-dihydro-6H-quinolin-5-one

  • MDL number MFCD08692066
  • Cas Number : 124467-36-3
  • Molecular Weight : 181.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart