LIF100873200 Life Chemicals Inc.

3-tert-butyl-1,2-oxazol-5-amine

  • MDL number MFCD05266697
  • Cas Number : 59669-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart