LIF100873271 Life Chemicals Inc.

6-Methyl-imidazo[2,1-b]thiazole-5-carbaldehyde

  • MDL number MFCD02253172
  • Cas Number : 75001-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart