LIF100873273 Life Chemicals Inc.

7-Bromo-benzo[1,3]dioxole-5-carbaldehyde

  • MDL number MFCD00128258
  • Cas Number : 19522-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart