LIF112841939 Life Chemicals Inc.

Methyl 4-(piperazin-1-ylsulfonyl)benzoate hydrochloride

  • MDL number MFCD09033845
  • Cas Number : 1177346-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart