LIF112842031 Life Chemicals Inc.

6-Pyridin-3-yl-2H-pyridazin-3-one

  • MDL number MFCD09033893
  • Cas Number : 78784-65-3
  • Molecular Weight : 173.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart