LIF112842046 Life Chemicals Inc.

1-(2-Fluorobenzyl)-2-oxo-1,2-dihydropyridine-3-carboxylic acid

  • MDL number MFCD09033901
  • Cas Number : 941869-31-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart