LIF112842047 Life Chemicals Inc.

1-(3,5-Dimethoxybenzyl)-2-oxo-1,2-dihydropyridine-3-carboxylic acid

  • MDL number MFCD09033902
  • Cas Number : 941869-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart