LIF112842657 Life Chemicals Inc.

1-(1H-Benzimidazol-2-yl)propan-1-ol

  • MDL number MFCD08163406
  • Cas Number : 4857-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart