LIF112842673 Life Chemicals Inc.

3',5'-dimethyl-1'-propyl-1H,1'H-3,4'-bipyrazole-5-carboxylic acid

  • MDL number MFCD07397888
  • Cas Number : 890621-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart