LIF112842928 Life Chemicals Inc.

1H-1,2,3-Benzotriazol-1-ylacetic acid

  • MDL number MFCD00553342
  • Cas Number : 4144-64-3
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart