LIF112850146 Life Chemicals Inc.

2-(1H-1,3-benzodiazol-1-yl)acetic acid hydrochloride

  • MDL number MFCD08163106
  • Cas Number : 936348-97-9
  • Molecular Weight : 212.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1H-benzimidazol-1-ylacetic acid hydrochloride
Inchi Code 1S/C9H8N2O2.ClH/c12-9(13)5-11-6-10-7-3-1-2-4-8(7)11;/h1-4,6H,5H2,(H,12,13);1H
Inchi Key VOWNVGNQMOICBJ-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart