LIF112851525 Life Chemicals Inc.

1'-Isopropyl-3',5'-dimethyl-1H,1'H-3,4'-bipyrazole-5-carboxylic acid

  • MDL number MFCD07791993
  • Cas Number : 1044275-20-8
  • Molecular Weight : 248.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1'-isopropyl-3',5'-dimethyl-1H,1'H-3,4'-bipyrazole-5-carboxylic acid
Inchi Code 1S/C12H16N4O2/c1-6(2)16-8(4)11(7(3)15-16)9-5-10(12(17)18)14-13-9/h5-6H,1-4H3,(H,13,14)(H,17,18)
Inchi Key YLRUIZVZRUFWDV-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart