LIF112851529 Life Chemicals Inc.

5-Methyl-4,5,6,7-tetrahydro-1H-indazole-3-carboxylic acid

  • MDL number MFCD03834488
  • Cas Number : 696645-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart