LIF112851542 Life Chemicals Inc.

2-Piperazin-1-yl-quinoline

  • MDL number MFCD00242605
  • Cas Number : 4774-24-7
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart